Skip to Content

C-14027

Human Pulmonary Microvascular Endothelial Cells (HPMEC)

1 million cells per pellet

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.81
UNSPSC Code:
41106514
Biological source:
human lung
Morphology:
(endothelial)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human lung

packaging

pkg of 1,000,000 cells

morphology

(endothelial)

application(s)

cell analysis (DNA, RNA, and protein)

shipped in

dry ice

storage temp.

−20°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details. Cells are dissolved in RNAlater for subsequent RNA, DNA, or protein analysis. Note that cell pellets cannot be revived.

Application

Primary Human Pulmonary Microvascular Endothelial Cells (HPMEC) are isolated from the lung from a single donor. Since lung tissue contains blood and lymphatic capillaries, HPMEC comprise Blood and Lymphatic Microvascular Endothelial Cells. The cells are routinely analyzed by immunofluorescent staining: they stain positive for CD31 and von Willebrand factor and negative for smooth muscle alpha-actin.The lung has a vast endothelial surface area, which is essential for the exchange of gases. HPMEC are most appropriate for studying human lung diseases.

Biochem/physiol Actions

Lungs

Physical form

Pellet is dissolved in RNAlater® for downstream analysis of DNA, RNA or protein profiles. Cells are not revivable.

Preparation Note

Click here for more information.

Analysis Note

Rigid quality control tests are performed for each lot of the corresponding cells. They are tested for cell viability, cell morphology, growth performance and for cell-type specific markers. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Other Notes

The Cell Pellets can be stored indefinitely at -20°C, and for up to 1 month at 4°C. Note: The Cell Pellets will not freeze at -20°C. Always centrifuge product briefly before opening vial.

Legal Information

RNAlater is a registered trademark of Thermo Fisher Scientific, Inc.

Disclaimer

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library