Skip to Content
MilliporeSigma
All Photos(1)

Documents

C0735

Sigma-Aldrich

8-(4-Chlorophenylthio)adenosine-3′,5′-cyclic Monophosphorothioate, Rp-isomer sodium salt

≥99% (HPLC), solid

Synonym(s):

Rp-8-CPT-cAMPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14ClN5O5PS2Na
CAS Number:
Molecular Weight:
509.86
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥99% (HPLC)

form

solid

packaging

vial of 5 μmol

storage temp.

−20°C

SMILES string

[Na+].Nc1ncnc2n([C@@H]3O[C@@H]4COP([S-])(=O)O[C@H]4[C@H]3O)c(Sc5ccc(Cl)cc5)nc12

InChI

1S/C16H15ClN5O5PS2.Na/c17-7-1-3-8(4-2-7)30-16-21-10-13(18)19-6-20-14(10)22(16)15-11(23)12-9(26-15)5-25-28(24,29)27-12;/h1-4,6,9,11-12,15,23H,5H2,(H,24,29)(H2,18,19,20);/q;+1/p-1/t9-,11-,12-,15-,28?;/m1./s1

InChI key

AWXMSJRRXLCVMW-JVSRKQJHSA-M

Biochem/physiol Actions

Lipophilic analog of the competitive inhibitor of protein kinase A (cyclic AMP antagonist).

Features and Benefits

This compound is featured on the PKA & PKG page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M G Weisskopf et al.
Science (New York, N.Y.), 265(5180), 1878-1882 (1994-09-23)
Repetitive activation of hippocampal mossy fibers evokes a long-term potentiation (LTP) of synaptic responses in pyramidal cells in the CA3 region that is independent of N-methyl-D-aspartate receptor activation. Previous results suggest that the site for both the induction and expression
Ting Zhou et al.
Journal of molecular neuroscience : MN, 68(1), 111-119 (2019-03-11)
Studies have shown that papaverine can inhibit lipopolysaccharide (LPS)-induced microglial activation. The retinal primary microglia of newborn SD rats were isolated and purified, and a LPS-induced microglia activation model was established. The protein phosphorylation level of the signaling pathway was
Chad M Estep et al.
PloS one, 11(12), e0169044-e0169044 (2016-12-31)
Dopaminergic (DA) neurons in the substantia nigra pars compacta (SNc) are richly innervated by GABAergic neurons. The postsynaptic effects of GABA on SNc DA neurons are mediated by a mixture of GABAA and GABAB receptors. Although activation of GABAA receptors
Dongqin Yang et al.
EBioMedicine, 47, 114-127 (2019-09-10)
There are many reports of the anti-tumour effects of exogenous adenosine in gastrointestinal tumours. Gemcitabine, a first line agent for patients with poor performance status, and adenosine have structural similarities. For these reasons, it is worth exploring the therapeutic efficacy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service