Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C0730

Sigma-Aldrich

Anti-CD137 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-4-1BB

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

capture ELISA: 0.2-0.8 μg/mL
flow cytometry: 2.5 μg using 106 human T cells treated with PHA
immunocytochemistry: 5-15 μg/mL
western blot: 0.1 μg/mL

impurities

<0.10 EU/μg endotoxin

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNFRSF9(3604)

General description

CD137 (4-1BB) is a member of the tumor necrosis factor (TNF) receptor superfamily. It is a type I transmembrane glycoprotein expressed on the cell surface of activated splenic T cells and thymocytes.

Immunogen

mouse myeloma cell line NS0-derived recombinant human 41BB/TNFRSF9/CD137 Leu24Gln186

Application

Anti-CD137 antibody produced in goat is suitable for the following applications:
  • Agonist Activity: Measured by its ability to costimulate IFNg secretion by human T cells in the presence of anti-CD3. The ED50 for this effect is typically 5-20μg/mL.
  • ELISA capture: It can be used as a capturing reagent in a human 4-1BB sandwich immunoassay in combination with biotinylated human 4-1BB detection antibody and recombinant human 4-1BB as the standard. The suggested coating concentration range is 0.2-0.8μg/ml. The recommended concentration of the detection antibody is 0.1-0.4μg/mL.
  • Flow cytometry: 2.5μg/106 cells using Human T cells treated with PHA (phytohemagglutinin).
  • Immunoblotting: A working concentration of 0.1μg/ml was determined using recombinant human CD137.
  • Immunocytochemistry: 5-15μg/mL using immersion fixed human peripheral blood mononuclear cells.

Biochem/physiol Actions

CD137 in T lymphocytes helps to regulate activation, proliferation and apoptosis. CD137 and CD28 are co-stimulatory molecules of T cell activation. It plays an important role in regulating T-cell-dependent immune responses. The expression of CD137 correlates negatively with lymphocyte proliferation and induces cell death caused by mitogen overstimulation. In monocytes, CD137 induces activation, promotes adherence and helps in prolongs survival.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Reconstitution

Reconstitute at 0.2 mg/mL in sterile PBS

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Kienzle et al.
International immunology, 12(1), 73-82 (1999-12-23)
Human CD137 is a member of the tumor necrosis factor (TNF) receptor family and the homologue of murine 4-1BB. Recent studies have demonstrated that CD137 promotes accessory T cell activation, and regulates proliferation and survival of T lymphocytes. This study
J Langstein et al.
Journal of leukocyte biology, 65(6), 829-833 (1999-06-25)
CD137 (ILA/4-1BB), a member of the tumor necrosis factor (TNF) receptor family, promotes adherence and prolongs survival of human peripheral monocytes. It induces a strong expression of macrophage colony-stimulating factor (M-CSF), an essential monocyte survival factor. Monocyte survival induced by
H H Kim et al.
Molecules and cells, 10(3), 247-252 (2000-07-20)
It has been widely accepted that T cell activation requires two signals; one from the binding of the antigen/major histocompatibility complex to the T-cell receptor (TCR)/CD3 complex and the other from the interaction between a surface molecule on antigen presenting
J C Hurtado et al.
Journal of immunology (Baltimore, Md. : 1950), 155(7), 3360-3367 (1995-10-01)
The expression of the murine T cell Ag 4-1BB, a member of the TNF-R family, is induced by T cell activation. Previously, we and others had shown that signaling through 4-1BB enhanced proliferative T cell responses. To investigate a potential
J Michel et al.
Cytokine, 12(6), 742-746 (2000-06-14)
CD137 is a member of the tumour necrosis factor receptor family which delivers a potent co-stimulatory signal to T lymphocytes. Soluble forms of CD137 (sCD137) can be found at enhanced levels in sera of patients with rheumatoid arthritis. Here we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service