Key Documents
BMSIPD960035ZO
Sigma-Aldrich
BeadBlaster 96 Grinding Jars for Trace Metal Samples
(zirconium oxide), capacity 35 mL, pack of 2
About This Item
UNSPSC Code:
41100000
NACRES:
NA.82
material
(zirconium oxide)
packaging
pack of 2
manufacturer/tradename
Benchmark Scientific IPD9600-35ZO
capacity
35 mL
compatibility
BeadBlaster 96
General description
The jars feature threaded closures for leakproof performance with both solids and liquids.
Zirconium oxide is a chemically inert ceramic material. Not as hard as and less expensive than tungsten carbide, it is used for softer samples where trace metals from stainless steel may be a concern. These jars are ideal for grinding biological samples, plants, bones, teeth, soils and sludges. Jars are offered in a 35ml volume.
Legal Information
BeadBlaster is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)