About This Item
NACRES:
NB.22
UNSPSC Code:
41104000
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37862-0002
General description
Provides a cost effective and accurate way to count colonies of bacteria or other microorganisms grown on an agar plate. This hand-held, simple to operate device marks, counts, and gives an audible and visual alert each time a count is registered. The lightweight, pen-style design reduces the likelihood of error, combining all aspects of counting into one motion. CE approved.
Features and Benefits
- Fast, accurate and economical sample counting and sample count verification for up to 35 dishes
- Ability to count both up and down to verify sample counts
- Can display the sum total of all colonies counted
- Backlit display allows easy viewing in low light conditions
- Audible alert can be turned off if desired
- Counting function can be locked to prevent error during breaks between counting
- Long life, red and black felt-tip markers (included) work on plastic or glass and are oil and water resistant; ink is removable with ethyl alcohol
- Includes 3V lithium battery and protective storage case
Legal Information
Scienceware is a registered trademark of Bel-Art Products, a division of SP
Choose from one of the most recent versions:
Already Own This Product?
Global Trade Item Number
| SKU | GTIN |
|---|---|
| BAF378620002-1EA | 04061832872209 |