Skip to Content
MilliporeSigma

B8407

Nalgene® narrow mouth PPCO bottle

polypropylene copolymer, capacity 250 mL, translucent, with cap

Synonym(s):

nalgene bottle, nalgene bottles, Nalgene 2006-0008, laboratory container, receptical, vessel

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.17
UNSPSC Code:
41121800
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene copolymer bottle (PPCO), round bottle, translucent , polypropylene copolymer

joint

Joints threaded neck (24)

description

Autoclavable

feature

with cap

packaging

pack of 12 ea

manufacturer/tradename

Nalgene® 2006-0008

bottle diam. × H

61 mm × 133 mm

capacity

250 mL

General description

For lab applications requiring excellent chemical resistance and autoclaving, with and without contents, use Nalgene®arrow-mouth PPCO bottles with Closure. These polypropylene copolymer bottles are translucent, providing good clarity for viewing contents. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables.

Features and Benefits

  • Nalgene® narrow-mouth bottles are ideal for long-term storage.
  • Made from polypropylene copolymer (PPCO) material for excellent sample clarity.
  • Highly chemical resistant to most acids, bases, and alcohols.
  • Autoclavable - with and without contents.
  • Laboratory-grade plastic materials for dependably low leachables and extractables.
  • Liner less caps ensures leak-proof seal for sample protection.
  • Light-weight - enables easy lifting and transporting.
  • Avoid UV exposure.
  • Manufactured from non-toxic materials.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?

Explore all of our products under Nalgene® narrow mouth PPCO bottle




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
B8407-12EA04061832613949