Skip to Content
MilliporeSigma

B800-05

Bovine Brain Artery Endothelial Cells: BBAEC (Cryovial)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.81
UNSPSC Code:
41106514
Biological source:
bovine brain (arteries)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine brain (arteries)

manufacturer/tradename

Cell Applications, Inc

karyotype

2n = 60

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Biochem/physiol Actions

Artery

Preparation Note

Please refer to the BBAEC Culture Protocol.


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

No data available

flash_point_c

No data available



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library