MilliporeSigma
B7910
Nalgene® bottles, style 2126
capacity 4000 mL
Synonym(s):
nalgene bottle, nalgene bottles, Nalgene 2126-4000, laboratory container, receptical, vessel
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.17
UNSPSC Code:
41121800
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polypropylene cap, polypropylene copolymer bottle (PPCO), round translucent bottle, white polypropylene screw closure
joint
Joints threaded neck (83B)
description
Autoclavable
feature
with cap
packaging
pack of 1 ea
manufacturer/tradename
Nalgene® 2126-4000
bottle diam. × H
155 mm × 338 mm
capacity
4000 mL
General description
Round, heavy-duty, thick walled, wide-mouth bottles. Screw closure contains a thermoplastic elastomer (TPE) gasket.
Features and Benefits
  • Nalgene® wide-mouth bottles are ideally used as a waste aspirator bottle or as an autoclavable vessel for scale-up activities in cell culture applications.
  • Made of heavy-duty chemically resistant polypropylene copolymer
  • Thick wall, can withstand repeated room-temperature vacuum applications.
  • A leak-proof closure prevents sample loss.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Still not finding the right product?
Explore all of our products under Nalgene® bottles, style 2126
Choose from one of the most recent versions:
Certificates of Analysis (COA)