Skip to Content
MilliporeSigma

Skip To

B7542

Maleimide Activated BSA

Synonym(s):

Maleimide Activated Bovine Serum Albumin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.32
UNSPSC Code:
12352111
Biological source:
bovine
Form:
lyophilized powder
Technique(s):
ELISA: suitable, bioconjugation: suitable
Concentration:
5—6 mg/mL protein
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

Quality Segment

sterility

non-sterile

form

lyophilized powder

concentration

5—6 mg/mL protein

technique(s)

ELISA: suitable, bioconjugation: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

bovine ... ALB(280717)

General description

Maleimide Activated BSA is provided in lyophilized, ready to-use form, eliminating the need for time-consuming activation and purification steps of the carrier protein. Maleimide Activated BSA contains approximately 5-15 maleimide groups for each BSA molecule.. The conjugation reaction with a cysteine containing peptide is performed at pH 6.6-7.0. The extent of conjugation can then be determined by a colorimetric assay with Ellman′s reagent) to detect unreacted thiol groups in the cysteine-containing peptide.

Application

Maleimide Activated BSA has been used in peptide conjugation.
Maleimide-activated BSA (mBSA) has reactive maleimide groups on its surface available for conjugation with a cysteine-containing peptide or a thiol-containing hapten. The reaction of the maleimide group and the thiol group proceeds rapidly and selectively under mild coupling conditions (pH 6.5-7.5) to yield a stable, covalently linked peptide-protein conjugate that may be used in immunization protocols or in antibody screening methods.

Physical form

Each vial contains 5 mg of Maleimide Activated BSA lyophilized from 10 mM sodium phosphate buffer, pH 6.6, with 0.115 mM NaCl, 1 mM EDTA, and 40 mM sucrose as stabilizer.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A3059A8022A1933
description

-

description

heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%

description

heat shock fraction, pH 5.2, ≥96%

description

chromatographically purified, New Zealand origin, low endotoxin, suitable for cell culture, pH 7, ≥98%

biological source

bovine

biological source

bovine

biological source

bovine

biological source

bovine

technique(s)

ELISA: suitable, bioconjugation: suitable

technique(s)

western blot: suitable, ELISA: suitable

technique(s)

western blot: suitable, ELISA: suitable, electrophoresis: suitable, blocking: suitable

technique(s)

cell culture | mammalian: suitable

Quality Level

200

Quality Level

300

Quality Level

300

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

sterility

non-sterile

sterility

-

sterility

-

sterility

-

Gene Information

bovine ... ALB(280717)

Gene Information

bovine ... ALB(280717)

Gene Information

bovine ... ALB(280717)

Gene Information

bovine ... ALB(280717)


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters