Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

B6756

Sigma-Aldrich

Brilliant Green

Dye content ~90 %, Powder

Synonym(s):

Astradiamant green GX, Basic Green 1, Diamond Green, Emerald Green, Ethyl Green, Malachite Green G, Solid Green JO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H34N2O4S
CAS Number:
Molecular Weight:
482.63
Colour Index Number:
42040
Beilstein:
3901207
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Brilliant Green, Dye content ≥85 %

form

powder

composition

Dye content, ≥85%

color

blue-green

mp

210 °C (dec.) (lit.)

solubility

H2O: 1 mg/mL

εmax

≥1750 at 628- 632  nm in 50% ethanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

OS([O-])(=O)=O.CCN(CC)c1ccc(cc1)C(\c2ccccc2)=C3/C=CC(\C=C3)=[N+](/CC)CC

InChI

1S/C27H33N2.H2O4S/c1-5-28(6-2)25-18-14-23(15-19-25)27(22-12-10-9-11-13-22)24-16-20-26(21-17-24)29(7-3)8-4;1-5(2,3)4/h9-21H,5-8H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

NNBFNNNWANBMTI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Brilliant Green has been used for the staining of nodule initials in Lupinus angustifolius and Arachis hypogaea.

Biochem/physiol Actions

Brilliant Green is a dye and an antiseptic agent.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of iron in nodulation and nitrogen fixation in Lupinus angustifolius L.
Caixian T, et al.
The New phytologist, 114, 173-173 (1990)
Juo PS
Concise Dictionary of Biomedicine and Molecular Biology (2001)
Iron-deficiency specifically limits nodule development in peanut inoculated with Bradyrhizobium sp.
O'HARA GW, et al.
The New phytologist, 108, 51-51 (1988)
T Park et al.
Journal of applied microbiology, 127(3), 670-682 (2019-06-06)
Experiments were designed to determine the effects of different chemical inhibitors of lysozyme and peptidases on rumen protozoa and the associated prokaryotes, and in vitro fermentation using Entodinium caudatum as a model protozoan species. Imidazole (a lysozyme inhibitor), phenylmethylsulphonyl fluoride
B K Nandi et al.
Journal of hazardous materials, 161(1), 387-395 (2008-05-06)
Experimental investigations were carried out to adsorb toxic brilliant green dye from aqueous medium using kaolin as an adsorbent. Characterization of kaolin is done by measuring: (i) particle size distribution using particle size analyzer, (ii) BET surface area using BET

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service