Skip to Content
MilliporeSigma
All Photos(2)

Documents

B5386

Sigma-Aldrich

8-Bromoadenosine 3′,5′-cyclic monophosphate

≥97% (HPLC), powder, protein kinase A activator

Synonym(s):

8-Br-A-3:5-MP, 8-Br-cAMP, 8-Bromo-cAMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11BrN5O6P
CAS Number:
Molecular Weight:
408.10
Beilstein:
591930
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

product name

8-Bromoadenosine 3′,5′-cyclic monophosphate, ≥97% (HPLC)

Quality Level

Assay

≥97% (HPLC)

form

powder

mp

254 °C (dec.) (lit.)

solubility

aqueous base: soluble, clear

storage temp.

−20°C

SMILES string

Nc1ncnc2n([C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O)c(Br)nc12

InChI

1S/C10H11BrN5O6P/c11-10-15-4-7(12)13-2-14-8(4)16(10)9-5(17)6-3(21-9)1-20-23(18,19)22-6/h2-3,5-6,9,17H,1H2,(H,18,19)(H2,12,13,14)/t3-,5-,6-,9-/m1/s1

InChI key

DVKQVRZMKBDMDH-UUOKFMHZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

8-Bromoadenosine 3′,5′-cyclic monophosphate has been used:
  • to treat H295R cells as positive controls for CYP19 induction
  • as a membrane permeable cAMP analog to study its effect on short-circuit current (Isc)
  • to investigate its potential as an inducer of differentiation in Wharton′s jelly-derived mesenchymal stem cells (WJ-MSCs)

Biochem/physiol Actions

8-Bromoadenosine 3′,5′-cyclic monophosphate is a cell-permeable cAMP analog having greater resistance to hydrolysis by phosphodiesterases than cAMP. 8-Bromoadenosine 3′,5′-cyclic monophosphate activates protein kinase A, inhibits growth, decreases proliferation, increases differentiation, and induces apoptosis of cultured cells.
8-bromo-adenosine 3′,5′-cyclic monophosphate (8-bromo-cAMP), added to hepatocytes during plating helps to increase the acquisition of beta-adrenoceptors.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

8-Bromoadenosine 3′,5′-cyclic monophosphate is a membrane-permeable cAMP analog.
Sensitive to light and moisture

Caution

Sensitive to light and moisture.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dopamine increases Na+ absorption in the Reissner's membrane of the gerbil cochlea
Kim C H, et al.
Auris, Nasus, Larynx, 40(3), 266-272 (2013)
8-Bromo-cAMP and 8-CPT-cAMP Increase the Density of beta-Adrenoceptors in Hepatocytes by a Mechanism Not Mimicking the Effect of cAMP
Sandnes D, et al.
Pharmacology & Toxicology, 79(1), 15-22 (1996)
Differentiation of human umbilical cord Wharton?s jelly-derived mesenchymal stem cells into endometrial cells
Shi Q, et al.
Stem Cell Research & Therapy, 8(1), 246-246 (2017)
2, 3, 7, 8-Tetrachlorodibenzo-p-dioxin and diindolylmethanes differentially induce cytochrome P450 1A1, 1B1, and 19 in H295R human adrenocortical carcinoma cells
Sanderson J T, et al.
Toxicological Sciences, 61(1), 40-48 (2001)
Akihiro Goto et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(11), 4901-4912 (2013-03-15)
Enteric neural crest-derived cells (ENCCs) migrate from the anterior foregut in a rostrocaudal direction to colonize the entire gastrointestinal tract and to form the enteric nervous system. Genetic approaches have identified many signaling molecules regulating the migration of ENCCs; however

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service