Skip to Content
MilliporeSigma

B4770

Buffer, reference standard

pH 7.00±0.01 (25 °C)

Synonym(s):

pH calibration solution at 25 °C pH 7.00±0.01

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12161700
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

color

color-coded yellow

pH

7.00±0.01 (25 °C)

Packaging

500 mL size packaged in polyethylene bottle; 1 gal size in convenient shelf-top dispenser with spigot.

Analysis Note

Standardized against N.I.S.T. standard reference materials.


Still not finding the right product?

Explore all of our products under Buffer, reference standard


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library