Skip to Content
MilliporeSigma

B3412

Berberine hemisulfate salt

≥95%

Synonym(s):

Natural Yellow 18

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C20H18NO4 · 1/2SO4
CAS Number:
Molecular Weight:
384.39
UNSPSC Code:
12171500
NACRES:
NA.32
PubChem Substance ID:
MDL number:
Colour Index Number:
75160
Assay:
≥95%
Form:
powder
Solubility:
methanol: soluble
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95%

form

powder

solubility

methanol: soluble

SMILES string

[O-]S([O-])(=O)=O.COc1ccc2cc3-c4cc5OCOc5cc4CC[n+]3cc2c1OC.COc6ccc7cc8-c9cc%10OCOc%10cc9CC[n+]8cc7c6OC

InChI

1S/2C20H18NO4.H2O4S/c2*1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2;1-5(2,3)4/h2*3-4,7-10H,5-6,11H2,1-2H3;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

OJVABJMSSDUECT-UHFFFAOYSA-L

Application

Fluorescent stain for heparin in mast cells

Biochem/physiol Actions

An alkaloid with weak antibiotic properties. Substrate for MDR efflux pumps. Antimicrobial activities of berberine is potentiated by the MDR inhibitor 5´-methoxyhydnocarpin (5´-MHC). Berberine upregulates the expression of Pgp in hepatoma cells. Treatment with berberine potentially results in the reduced accumulation of chemotherapeutic drugs.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library