Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYT1000BRS

Pipette tips

racked, capacity 1000 μL, blue, pkg of 5x1000tips/cs, sterile

Synonym(s):

Pipette tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
1000 μL
sterility:
sterile
feature:
DNA free
RNase free
barrier: no
endotoxin free
filter: no
packaging:
pkg of 5x1000tips/cs
manufacturer/tradename:
Axygen Inc T-1000-B-R-S
Corning

material

blue

sterility

sterile

feature

DNA free
RNase free
barrier: no
endotoxin free
filter: no

manufacturer/tradename

Axygen Inc T-1000-B-R-S
Corning

packaging

pkg of 5x1000tips/cs

capacity

1000 μL

volume

1000 μL

color

blue

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

  • The Axygen® pipette tips are made of polypropylene.
  • The tips are optimized for use with both single and multi-channel pipettors.
  • Features a flexible wall and internal sealing ring for a secure fit.
  • The beveled tips reduce the capillary effect and facilitate accurate dispensing of the samples.

Other Notes

Browse all pipette tips

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service