Skip to Content
MilliporeSigma

AXYST45

Screw cap tubes without caps, conical bottom

size 0.5 mL, clear, pkg of 8x500caps/cs

Synonym(s):

centrifuge tubes

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.11
UNSPSC Code:
41121800
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Axygen Inc ST-45, Corning

packaging

pkg of 8x500caps/cs

size

0.5 mL

color

clear



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library