Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYPCR96LPFLTW

PCR microplate with flat top

low profile, size 96 wells, pkg of 4x25plates/cs, white

Synonym(s):

96 multiwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

material

conical bottom wells

manufacturer/tradename

Axygen Inc PCR-96-LP-FLT-W
Corning

packaging

pkg of 4x25plates/cs

size

96 wells

color

white

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service