Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYPCR384M2C

PCR microplate for ABI 3730, 3100 and MJ Thermocyclers

size 384 wells, Compatible with ABI, Full Skirt, clear, conical bottom clear polypropylene wells, pkg of 5x10plates/cs

Synonym(s):

384 multiwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

material

conical bottom clear polypropylene wells

sterility

non-sterile

feature

lid: no
skirt Full skirt

manufacturer/tradename

Axygen Inc PCR-384M2-C
Corning

packaging

pkg of 5x10plates/cs

maximum volume

50 μL

size

384 wells

working volume

50 μL

color

clear

suitability

suitable for (qPCR)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service