Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYPCR384LC480W

PCR microplate for Roche® 480 LightCycler®

with film, size 384 wells, white, pkg of 5x10plates/cs, conical bottom white polypropylene wells

Synonym(s):

96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
conical bottom white polypropylene wells
size:
384 wells
Pricing and availability is not currently available.

material

conical bottom white polypropylene wells

sterility

non-sterile

manufacturer/tradename

Axygen Inc PCR-384-LC480-W
Corning

packaging

pkg of 5x10plates/cs

maximum volume

30 μL

size

384 wells

working volume

30 μL

color

white

suitability

suitable for PCR

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Axygen® PCR microplate is ideal for use with Roche Light Cycler 480 system.
  • Uniform ultra-thin wall thickness facilitates optimal thermal transfer.
  • Includes Ultra Clear sealing film.
  • A printed alphanumeric grid enables well identification.
  • Free of DNase-/RNase and nonpyrogenic.

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
LightCycler is a registered trademark of Roche
Roche is a registered trademark of Roche Molecular Systems, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service