Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYPCR0208CPC

PCR strip tubes

with strip caps, thin wall, 8 per strip, size 0.2 mL, clear, pkg of 10x125strips/cs

Synonym(s):

96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

material

cap (domed strip caps)
polypropylene

sterility

non-sterile

manufacturer/tradename

Axygen Inc PCR-0208-CP-C
Corning

packaging

pkg of 10x125strips/cs

size

0.2 mL

color

clear
clear

suitability

suitable for PCR

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Made up of virgin polypropylene
  • Allows precise thermal transfer due to ultra-thin and consistent wall thickness
  • To prevent evaporation cap ensures a tight fit
  • Free From RNase-/DNase and nonpyrogenic in nature
  • Compatible with a wide variety of thermal cyclers

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service