Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYMTX1250CR

Matrix pipette tips

racked, capacity 1250 μL, pkg of 3x1152tips/cs

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
capacity:
1250 μL
packaging:
pkg of 3x1152tips/cs
manufacturer/tradename:
Axygen Inc MTX-1250-C-R
Corning

manufacturer/tradename

Axygen Inc MTX-1250-C-R
Corning

packaging

pkg of 3x1152tips/cs

capacity

1250 μL

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service