Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYMCT175LC

MAXYMum Recovery® microtubes

size 1.7 mL, pkg of 10x250tubes/cs

Synonym(s):

centrifuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121702
NACRES:
NB.22

material

cap (snap cap)
conical bottom tube
polypropylene
polypropylene cap

sterility

non-sterile

feature

RNase and DNase free
endotoxin free
graduations

manufacturer/tradename

Axygen Inc MCT-175-L-C
Corning

packaging

pkg of 10x250tubes/cs

size

1.7 mL

color

clear

fitting

cap (push fit)

suitability

suitable for (centrifugation)

binding type

(ultra-low retention)

Looking for similar products? Visit Product Comparison Guide

Legal Information

Maxymum Recovery is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service