Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV54586

Sigma-Aldrich

Anti-ACADSB antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-2-MEBCAD, Anti-ACAD7, Anti-Acyl-coenzyme A dehydrogenase, short/branched chain, Anti-SBCAD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
human, rat, mouse, bovine
citations:
4
technique(s):
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

44 kDa

species reactivity

human, rat, mouse, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACADSB(36)

Immunogen

Synthetic peptide directed towards the middle region of human ACADSB

Application

Anti-ACADSB antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.

Biochem/physiol Actions

The protein encoded by ACADSB (Acyl-coenzyme A dehydrogenase, short/branched chain) gene belongs to acyl-CoA dehydrogenases (ACADs) family and is mapped on to chromosome 10 at 10q25-q26. It is a homotetramer with each monomer comprising of a non-covalently bound flavin adenine dinucleotide (FAD) molecule as a cofactor. It catalyzes the initial step of mitochondrial fatty acid β-oxidation for substrates with four and six carbons. ACADSB also catalyzes the third step of leucine and isoleucine/valine metabolism. Deficiency of the encoded protein increases 2-methylbutyrylglycine and 2-methylbutyrylcarnitine in blood and urine and results in catabolism of L-isoleucine.

Sequence

Synthetic peptide located within the following region: GLRASSTCPLTFENVKVPEANILGQIGHGYKYAIGSLNEGRIGIAAQMLG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Binzak et al.
Biochimica et biophysica acta, 1382(1), 137-142 (1998-03-21)
The acyl-CoA dehydrogenases (ACDs) are a family of related enzymes which catalyze the alpha,beta-dehydrogenation of acyl-CoA esters, transferring electrons to electron transferring flavoprotein. We have recently cloned and characterized the cDNA for human short/branched chain acyl-CoA dehydrogenase (SBCAD). Based on
Amy K Saenger et al.
Biochemistry, 44(49), 16043-16053 (2005-12-08)
Human short-chain acyl-CoA dehydrogenase (hSCAD) catalyzes the first matrix step in the mitochondrial beta-oxidation cycle for substrates with four and six carbons. Previous studies have shown that the act of substrate/product binding induces a large enzyme potential shift in acyl-CoA
K M Gibson et al.
Pediatric research, 47(6), 830-833 (2000-06-01)
An 4-mo-old male was found to have an isolated increase in 2-methylbutyrylglycine (2-MBG) and 2-methylbutyrylcamitine (2-MBC) in physiologic fluids. In vitro oxidation studies in cultured fibroblasts using 13C- and 14C-labeled branched chain amino acids indicated an isolated block in 2-methylbutyryl-CoA
P Deloukas et al.
Nature, 429(6990), 375-381 (2004-05-28)
The finished sequence of human chromosome 10 comprises a total of 131,666,441 base pairs. It represents 99.4% of the euchromatic DNA and includes one megabase of heterochromatic sequence within the pericentromeric region of the short and long arm of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service