Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV50153

Sigma-Aldrich

Anti-PCDH15 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DFNB23, Anti-DKFZp667A1711, Anti-Protocadherin 15, Anti-RP11-449J3.2, Anti-USH1F

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

80 kDa

species reactivity

human, guinea pig, rat, rabbit, mouse, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCDH15(65217)

Related Categories

General description

The gene Protocadherin-related 15 (PCDH15) is mapped to human chromosme 10q21.1. It is an integral membrane protein and one of the members of the cadherin superfamily that mediate calcium-dependent cell-cell adhesion.. PCDH15 transcripts can be detected in adult brain, lung, kidney, fetal retina and fetal cochlea.

Immunogen

Synthetic peptide directed towards the N terminal region of human PCDH15

Application

Anti-PCDH15 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

The expression of Protocadherin-related 15 (PCDH15) is essential for normal function of cochlea and retina. Mutations in this gene result in hearing loss and Usher Syndrome Type IF (USH1F). PCDH15 is expressed in nasal NK/T-cell lymphomas.

Sequence

Synthetic peptide located within the following region: HSIVVQVQCINKKVGTIIYHEVRIVVRDRNDNSPTFKHESYYATVNELTP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebeca Burgos-Panadero et al.
BMC cancer, 19(1), 479-479 (2019-05-24)
Vitronectin is a multifunctional glycoprotein known in several human tumors for its adhesive role in processes such as cell growth, angiogenesis and metastasis. In this study, we examined vitronectin expression in neuroblastoma to investigate whether this molecule takes part in
Eléonore Lambert et al.
Scientific reports, 8(1), 9837-9837 (2018-07-01)
Tetrastatin, a 230 amino acid sequence from collagen IV, was previously demonstrated to inhibit melanoma progression. In the present paper, we identified the minimal active sequence (QKISRCQVCVKYS: QS-13) that reproduced the anti-tumor effects of whole Tetrastatin in vivo and in
Yinjun Dong et al.
Molecular imaging and biology, 21(1), 175-182 (2018-06-06)
To assess a novel radiotracer aluminum [18F]fluoride-1,4,7-triazacyclononane-triacetic acid-pegylated dimeric RGD ([18F]ALF-NOTA-PRGD2, denoted as [18F]Alfatide) for positron emission tomography (PET)/X-ray computed tomography (CT) and explore the relationships between clinicopathological characteristics and maximum standard uptake values in primary (SUVP) and metastatic lymph
Jia Hou et al.
Science advances, 5(7), eaau8301-eaau8301 (2019-09-19)
Cerebral ischemia (CI) results from inadequate blood flow to the brain. The difficulty of delivering therapeutic molecules to lesions resulting from CI hinders the effective treatment of this disease. The inflammatory response following CI offers a unique opportunity for drug
V Rouget-Quermalet et al.
Oncogene, 25(19), 2807-2811 (2005-12-22)
Natural killer cells are well known to play an important role in immune defense against tumor development and viral infections. To further characterize new functionally relevant structures in these cells, we studied a series of monoclonal antibodies that we have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service