Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV48473

Sigma-Aldrich

Anti-MTR antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Mtr Antibody, Mtr Antibody - Anti-MTR antibody produced in rabbit, Anti-5-Methyltetrahydrofolate-homocysteine methyltransferase, Anti-FLJ45386

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

140 kDa

species reactivity

rat, guinea pig, mouse, horse, human, rabbit, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MTR(4548)

General description

MTR codes for 5-methyltetrahydrofolate-homocysteine methyltransferase that catalyzes the last step in methionine synthesis. Genetic alterations in MTR have been associated with methylcobalamin deficiency, breast cancer risk and prostate cancer susceptibility.
Rabbit Anti-MTR antibody recognizes chicken, human, mouse, rat, and bovine MTR.

Immunogen

Synthetic peptide directed towards the C terminal region of human MTR

Application

Rabbit Anti-MTR antibody is suitable for western blot applications at a concentration of 1μg/ml.

Biochem/physiol Actions

MTR is the enzyme 5-methyltetrahydrofolate-homocysteine methyltransferase. This enzyme, also known as cobalamin-dependent methionine synthase, catalyzes the final step in methionine biosynthesis. Mutations in MTR have been identified as the underlying cause of methylcobalamin deficiency complementation group G. MTR encodes the enzyme 5-methyltetrahydrofolate-homocysteine methyltransferase. This enzyme, also known as cobalamin-dependent methionine synthase, catalyzes the final step in methionine biosynthesis. Mutations in MTR have been identified as the underlying cause of methylcobalamin deficiency complementation group G. Sequence Note: This RefSeq record was created from transcript and genomic sequence data because no single transcript was available for the full length of the gene. The extent of this transcript is supported by transcript alignments. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: GSEQLDVADLRRLRYKGIRPAPGYPSQPDHTEKLTMWRLADIEQSTGIRL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrés López-Cortés et al.
The American journal of the medical sciences, 346(6), 447-454 (2013-03-06)
The methylenetetrahydrofolate reductase (MTHFR), methionine synthase (MTR) and MTR reductase (MTRR) enzymes act in the folate metabolism, which is essential in methylation and synthesis of nucleic acids. The single nucleotide polymorphisms, MTHFR C677T, A1298C, MTR A2756G and MTRR A66G, cause
Rita de Cássia Carvalho Barbosa et al.
Anticancer research, 32(11), 4805-4811 (2012-11-17)
Polymorphisms in genes encoding enzymes of folate metabolism are a focus of breast cancer risk studies due of the role of these enzymes in DNA methylation, synthesis, and repair. MTHFR, encoding for 5,10-methylenetetrahydrofolate reductase, is one of the most studied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service