Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV48247

Sigma-Aldrich

Anti-SEP15 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Selenoprotein 15 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

15 kDa

species reactivity

mouse, rat, pig, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SEP15(9403)

General description

SEP15 is a selenoprotein that regulates the unfolded protein response. It is modulated by ER stresses. SEP15 variations have been linked to the risk of lung cancer.
Rabbit Anti-SEP15 antibody recognizes bovine, zebrafish, pig, human, mouse, rat, and canine SEP15.

Immunogen

Synthetic peptide directed towards the middle region of human SEP15

Application

Rabbit Anti-SEP15 antibody is suitable for western blot applications at a concentration of 1μg/ml.

Biochem/physiol Actions

SEP15 is a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. Studies in mouse suggest that this selenoprotein may have redox function and may be involved in the quality control of protein folding. The gene that encodes the protein is localized on chromosome 1p31, a genetic locus commonly mutated or deleted in human cancers. This gene encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3′ UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Studies in mouse suggest that this selenoprotein may have redox function and may be involved in the quality control of protein folding. This gene is localized on chromosome 1p31, a genetic locus commonly mutated or deleted in human cancers. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.

Sequence

Synthetic peptide located within the following region: SDKPKLFRGLQIKYVRGSDPVLKLLDDNGNIAEELSILKWNTDSVEEFLS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vyacheslav M Labunskyy et al.
Biochemistry, 48(35), 8458-8465 (2009-08-05)
The accumulation of misfolded proteins in the endoplasmic reticulum (ER) results in activation of signaling pathways collectively known as the unfolded protein response (UPR). The UPR promotes adaptation of cells to ER stress by transient inhibition of protein translation and
Ewa Jablonska et al.
European journal of nutrition, 47(1), 47-54 (2008-02-02)
Selenium (Se) is a trace element suggested to act chemopreventive in lung cancer. The mechanism by which Se suppresses tumour development may be associated with some of the functions of selenoproteins, including 15 kDa selenoprotein (Sep15). This protein exhibits antioxidant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service