Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV48088

Sigma-Aldrich

Anti-PAX8 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Paired box gene 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

48 kDa

species reactivity

human, bovine, dog, mouse, rabbit, guinea pig, horse, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PAX8(7849)

General description

PAX8 is a nephric transcription factor that is involved in the embryogenesis of thyroid gland, renal, and Mullerian system. It is expressed in several ovarian and kidney carcinomas, and can be a useful biomarker for these cancers for determining primary tumor sites.
Rabbit Anti-PAX8 antibody recognizes zebrafish, human, mouse, rat, and canine PAX8.

Immunogen

Synthetic peptide directed towards the N terminal region of human PAX8

Application

Rabbit Anti-PAX8 antibody is suitable for western blot applications at a concentration of 0.25μg/ml.

Biochem/physiol Actions

PAX8 is a member of the paired box (PAX) family of transcription factors. Members of this family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. This nuclear protein is involved in thyroid follicular cell development and expression of thyroid-specific genes. Mutations in its gene have been associated with thyroid dysgenesis, thyroid follicular carcinomas and atypical follicular thyroid adenomas.This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically encode proteins which contain a paired box domain, an octapeptide, and a paired-type homeodomain. This nuclear protein is involved in thyroid follicular cell development and expression of thyroid-specific genes. Mutations in this gene have been associated with thyroid dysgenesis, thyroid follicular carcinomas and atypical follicular thyroid adenomas. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

Sequence

Synthetic peptide located within the following region: KSLSPGHTLIPSSAVTPPESPQSDSLGSTYSINGLLGIAQPGSDKRKMDD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prashant Kumar et al.
Genes & diseases, 9(1), 187-200 (2022-01-11)
TSC renal cystic disease is poorly understood and has no approved treatment. In a new principal cell-targeted murine model of Tsc cystic disease, the renal cystic epithelium is mostly composed of type A intercalated cells with an intact Tsc2 gene
David Tacha et al.
Applied immunohistochemistry & molecular morphology : AIMM, 19(4), 293-299 (2011-02-03)
PAX8 is a nephric-lineage transcription factor and is a crucial transcription factor for organogenesis of the thyroid gland, kidney, and Müllerian system. PAX8 is shown to be expressed in a high percentage of kidney and ovarian carcinomas. Limited information is
Anna R Laury et al.
The American journal of surgical pathology, 35(6), 816-826 (2011-05-10)
PAX8 is a paired-box gene important in embryogenesis of the thyroid, Müllerian, and renal/upper urinary tracts, and expression of PAX8 has been previously described in carcinomas from each of these sites. However, a large study including a wide variety of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service