Skip to Content
MilliporeSigma

AV46129

Anti-SHMT2 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-GLYA, Anti-SHMT, Anti-Serine hydroxymethyltransferase 2 (mitochondrial)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

56 kDa

species reactivity

horse, dog, rat, guinea pig, mouse, human, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SHMT2(6472)

Immunogen

Synthetic peptide directed towards the C terminal region of human SHMT2

Application

Anti-SHMT2 (AB1) antibody produced in rabbit is suitable for western blotting at a concentration of 0.5μg/ml and for immunohistochemistry at a concentration of 4-8μg/ml.

Biochem/physiol Actions

SHMT2 gene encodes an enzyme serine hydroxymethyltransferase 2 (mitochondrial), which is a pyridoxal phosphate-dependent enzyme that regulates the biosynthesis of thymidylate in mammals. It catalyzes the reversible reaction of serine and tetrahydrofolate to glycine and 5,10-methylene tetrahydrofolate. Additionally, it facilitates the interconversion of serine and glycine and is associated with mitochondrial DNA which assists in preventing the uracil accumulation in mtDNA.

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Other Notes

Synthetic peptide located within the following region: ELVSITANKNTCPGDRSAITPGGLRLGAPALTSRQFREDDFRRVVDFIDE

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library