Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV45680

Sigma-Aldrich

Anti-KNG1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-BDK, Anti-KNG, Anti-Kininogen 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

72 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KNG1(3827)

Related Categories

General description

Kininogen 1 (KNG1) a precursor of the kinin has recently been identified as possible biomarkers for chronic hepatitis C and proliferative vitreoretinopathy (PVR).

Specificity

Anti-KNG1 polyclonal antibody reacts with human and mouse kininogen 1 proteins.

Immunogen

Synthetic peptide directed towards the middle region of human KNG1

Application

Anti-KNG1 polyclonal antibody is used to tag kininogen 1 protein for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of kininogen 1 as a potential biomarker for chronic hepatitis C and proliferative vitreoretinopathy (PVR).

Biochem/physiol Actions

Kininogens are inhibitors of thiol proteases. HMW-kininogen plays an important role in blood coagulation by helping to position optimally prekallikrein and factor XI next to factor XII and inhibits the thrombin- and plasmin-induced aggregation of thrombocytes. The active peptide bradykinin that is released from HMW-kininogen shows a variety of physiological effects such as influence in smooth muscle contraction, induction of hypotension, natriuresis and diuresis, etc.

Sequence

Synthetic peptide located within the following region: YPGKDFVQPPTKICVGCPRDIPTNSPELEETLTHTITKLNAENNATFYFK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service