Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV44535

Sigma-Aldrich

Anti-SDCBP antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-MDA-9, Anti-ST1, Anti-SYCL, Anti-Syndecan binding protein (syntenin), Anti-TACIP18

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

32 kDa

species reactivity

human, rat, rabbit, guinea pig, horse, mouse, bovine, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SDCBP(6386)

Immunogen

Synthetic peptide directed towards the middle region of human SDCBP

Application

Anti-SDCBP antibody produced in rabbit is suitable for western blotting at a concentration of 1.25μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.

Biochem/physiol Actions

Syndecan binding protein (SDCBP; syntenin, mda-9) is a scaffolding protein containing a PDZ domain that has functions that include cell adhesion, proliferation, protein trafficking, cytoskeletal organization and activation of transcription factors. It promotes metastasis of cancer cells by activating FAK, NF-κB, c-Src and p38-MAPK pathways.

Sequence

Synthetic peptide located within the following region: VGIRRAEIKQGIREVILCKDQDGKIGLRLKSIDNGIFVQLVQANSPASLV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Habib Boukerche et al.
Cancer research, 67(4), 1812-1822 (2007-02-20)
mda-9/Syntenin is a scaffolding PDZ domain-containing protein overexpressed in multiple human cancers that functions as a positive regulator of melanoma metastasis. Using a normal immortal human melanocyte cell line and weakly and highly metastatic human melanoma cell lines, we presently
Xiao-Long Qian et al.
PloS one, 8(3), e60046-e60046 (2013-03-28)
Syndecan binding protein (SDCBP), an adapter protein containing PDZ domains, contributes to the tumorigenicity and metastasis of many malignant tumors, such as malignant melanoma. Our study aimed in revealing the expression profile of SDCBP in breast cancer (BCa) and its
Devanand Sarkar et al.
Cancer research, 68(9), 3087-3093 (2008-05-03)
Cancer is a progressive disease that, in many instances, if untreated, can culminate in metastatic spread of primary tumor cells to distant sites in the body. Metastasis frequently confers virulence and therapy resistance to cancer cells, and defining the molecular
Habib Boukerche et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(41), 15914-15919 (2008-10-04)
The scaffold PDZ-domain containing protein mda-9/syntenin functions as a positive regulator of cancer cell progression in human melanoma and other tumors. mda-9/Syntenin regulates cell motility and invasion by altering defined biochemical and signaling pathways, including focal adhesion kinase (FAK), p38

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service