Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV44161

Sigma-Aldrich

Anti-SLC6A18 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-FLJ31236, Anti-Solute carrier family 6, member 18, Anti-Xtrp2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
WB
clone:
polyclonal
species reactivity:
rat, human, mouse
citations:
2
technique(s):
immunohistochemistry: suitable
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

69 kDa

species reactivity

rat, human, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Synthetic peptide directed towards the middle region of human SLC6A18

Application

Anti-SLC6A18 antibody produced in rabbit is suitable for western blotting at a concentration of 1.25μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.

Biochem/physiol Actions

SLC6A18 (XT2) belongs to SLC6 family of transporters that mediate the movement of neurotransmitters, osmolytes and amino acids. XT2 is a neutral amino acid transporter present at the luminal membrane of kidney proximal tubules. The expression of SLC6A18 is associated with increased risk of myocardial infarction in a Japanese population.

Sequence

Synthetic peptide located within the following region: MHLNATWPKRVAQLPLKACLLEDFLDKSASGPGLAFVVFTETDLHMPGAP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dustin Singer et al.
The Journal of biological chemistry, 284(30), 19953-19960 (2009-05-30)
The orphan transporter Slc6a18 (XT2) is highly expressed at the luminal membrane of kidney proximal tubules and displays approximately 50% identity with Slc6a19 (B(0)AT1), which is the main neutral amino acid transporter in both kidney and small intestine. As yet
Ken Matsumoto et al.
Clinical biochemistry, 44(10-11), 789-794 (2011-03-23)
SLC6A18 (solute carrier family 6, member 18) acts as a specific transporter for neurotransmitters, amino acids and osmolytes such as betaine, taurine and creatine. The aim of the present study was to investigate the relationship between the human SLC6A18 gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service