Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV44116

Sigma-Aldrich

Anti-SLC43A2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ23848, Anti-LAT4, Anti-MGC34680, Anti-Solute carrier family 43, member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

53 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Solute carrier family 43, member 2 (SLC43A2, LAT4) is a system L amino acid transporter that preferentially transports L-tyrosine. Other members of the L amino acid transporter family include LAT1, LAT2 and LAT3. LAT4 may be an important amino acid transporter during gestation wherein it facilitates amino acid transport from the placenta into the fetus.

Specificity

Anti-SLC43A2 polyclonal antibody reacts with bovine, canine, rat, and human solute carrier family 43, member 2 proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human SLC43A2

Application

Anti-SLC43A2 polyclonal antibody is used to tag solute carrier family 43, member 2 for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of solute carrier family 43, member 2 in transport of tyrosine during fetal gestation.

Biochem/physiol Actions

SLC43A2 is a Sodium-, chloride, pH-independent high affinity transport of large neutral amino acids.

Sequence

Synthetic peptide located within the following region: TEPENVTNGTVGGTAEPGHEEVSWMNGWLSCQAQDEMLNLAFTVGSFLLS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service