Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV43340

Sigma-Aldrich

Anti-RNF146 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-DKFZP434O1427, Anti-RP3-351K20.1, Anti-Ring finger protein 146, Anti-dJ351K20.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39 kDa

species reactivity

mouse, horse, rat, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF146(81847)

General description

Ring finger protein 146 (RNF146, Iduna) is a poly(ADP-ribose) polymer binding and PARsylation dependent E3 ubiquitin ligase which ubiquitinates proteins for degradation by proteosomes. RNF146 is neuroprotective due to its inhibition of Parthanatos via binding with Poly(ADP-ribose) (PAR). RNF146 regulates Wnt signaling via ubiquitylation of auto-PARsylated tankyrase.

Specificity

Anti-RNF146 polyclonal antibody reacts with human and mouse ring finger protein 146 proteins.

Immunogen

Synthetic peptide directed towards the C terminal region of human RNF146

Application

Anti-RNF146 polyclonal antibody is used to tag ring finger protein 146 for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles ring finger protein 146 in Wnt signaling and neuroprotection.

Sequence

Synthetic peptide located within the following region: AVVAQHSLTQQRLLVSNANQTVPDRSDRSGTDRSVAGGGTVSVSVRSRRP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service