Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV41877

Sigma-Aldrich

Anti-CES1 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-ACAT, Anti-CEH, Anti-CES2, Anti-Carboxylesterase 1 (monocyte/macrophage Serine esterase 1), Anti-HMSE, Anti-HMSE1, Anti-MGC117365, Anti-PCE-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

61 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CES1(1066)

General description

Human monocyte serine esterase 1 (HMSE1) is an esterase isoenzyme expressed in monocyte/macrophages that can be inhibited by NaF in α-naphthyl acetate cytochemical staining applications. Intensive analysis of normal and malignant hematopoietic cells and leukemia-lymphoma cell lines using isoelectric focusing established the specificity of this enzyme for monocytes and related cells, hence its designation as monocyte-specific esterase. Monocyte-specific (carboxyl)esterase (MSE) is a marker enzyme which is well-known to hematologists as its detection is part of the traditional cytochemical stainings of monocyte lineage leukemia cells.

Specificity

Anti-CES1 (AB1) polyclonal antibody reacts with bovine, canine, rabbit, pig, human, mouse, and rat monocyte serine esterase 1 (MSE1) proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human CES1

Application

Anti-CES1 (AB1) polyclonal antibody is used to tag monocyte serine esterase 1 proteins for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the role of monocyte serine esterase 1 in monocytes and monocyte lineage leukemia.

Biochem/physiol Actions

CES1 is one of the enzymes responsible for the hydrolysis of ester- and amide-bond-containing drugs such as cocaine and heroin. They also hydrolize long-chain fatty acid esters and thioesters. This enzyme is known to hydrolyze aromatic and aliphatic esters and is necessary for cellular cholesterol esterification. It may also play a role in detoxification in the lung and/or protection of the central nervous system from ester or amide compounds. Carboxylesterase deficiency may be associated with non-Hodgkin lymphoma or B-cell lymphocytic leukemia.Carboxylesterase 1 is a member of a large multigene family. The enzymes encoded by these genes are responsible for the hydrolysis of ester- and amide-bond-containing drugs such as cocaine and heroin. They also hydrolize long-chain fatty acid esters and thioesters. This enzyme is known to hydrolyze aromatic and aliphatic esters and is necessary for cellular cholesterol esterification. It may also play a role in detoxification in the lung and/or protection of the central nervous system from ester or amide compounds. Carboxylesterase deficiency may be associated with non-Hodgkin lymphoma or B-cell lymphocytic leukemia. Three transcript variants encoding three different isoforms have been found for this gene.Carboxylesterase 1 is a member of a large multigene family. The enzymes encoded by these genes are responsible for the hydrolysis of ester- and amide-bond-containing drugs such as cocaine and heroin. They also hydrolize long-chain fatty acid esters and thioesters. This enzyme is known to hydrolyze aromatic and aliphatic esters and is necessary for cellular cholesterol esterification. It may also play a role in detoxification in the lung and/or protection of the central nervous system from ester or amide compounds. Carboxylesterase deficiency may be associated with non-Hodgkin lymphoma or B-cell lymphocytic leukemia. Three transcript variants encoding three different isoforms have been found for this gene.

Sequence

Synthetic peptide located within the following region: VLGKFVSLEGFAQPVAIFLGIPFAKPPLGPLRFTPPQPAEPWSFVKNATS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohit P Mathew et al.
Chembiochem : a European journal of chemical biology, 18(13), 1204-1215 (2017-02-22)
This report describes the metabolic glycoengineering (MGE) of intracellular esterase activity in human colon cancer (LS174T) and Chinese hamster ovary (CHO) cells. In silico analysis of carboxylesterases CES1 and CES2 suggested that these enzymes are modified with sialylated N-glycans, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service