Skip to Content
MilliporeSigma
All Photos(3)

Documents

AV40301

Sigma-Aldrich

Anti-RAE1 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-RAE1 RNA export 1 homolog (S. pombe)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

40 kDa

species reactivity

mouse, guinea pig, rabbit, human, dog, bovine, rat, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAE1(8480)

Related Categories

General description

RNA export 1 homolog (S. pombe) (RAE1) is involved in the nucleo-cytoplamic transport of mRNA. RAE1 has a novel postmitotic function in neural development via its interaction with RPM-1 (Regulator of Presynaptic Morphology-1) which regulates axon termination and synapse formation. RAE1 is a component of the Highwire (Hiw)/Drosophila Fsn E3 ubiquitin ligase complex required for normal synaptic morphology during development and axonal regeneration after injury. Rae1 restrains synaptic terminal growth by regulating the MAP kinase kinase kinase Wallenda. RNA export factor RAE1 contributes to NUP98-HOXA9-mediated leukemogenesis.

Specificity

Anti-RAE1 (AB1) polyclonal antibody reacts with zebrafish, bovine, canine, human, mouse, rat, chicken, and pig RNA export 1 homolog (S. pombe) proteins.

Immunogen

Synthetic peptide directed towards the C terminal region of human RAE1

Application

Anti-RAE1 (AB1) polyclonal antibody is used to tag RNA export 1 homolog (S. pombe) for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles RNA export 1 homolog (S. pombe) in nucleo-cytoplamic transport and the regulation of neural development and leukemogenesis.

Biochem/physiol Actions

RAE1 is a homolog of yeast Rae1. It contains four WD40 motifs, and has been shown to localize to distinct foci in the nucleoplasm, to the nuclear rim, and to meshwork-like structures throughout the cytoplasm. This gene is thought to be involved in nucleocytoplasmic transport, and in directly or indirectly attaching cytoplasmic mRNPs to the cytoskeleton.Mutations in the Schizosaccharomyces pombe Rae1 and Saccharomyces cerevisiae Gle2 genes have been shown to result in accumulation of poly(A)-containing mRNA in the nucleus, suggesting that the encoded proteins are involved in RNA export. The protein encoded by this gene is a homolog of yeast Rae1. It contains four WD40 motifs, and has been shown to localize to distinct foci in the nucleoplasm, to the nuclear rim, and to meshwork-like structures throughout the cytoplasm. This gene is thought to be involved in nucleocytoplasmic transport, and in directly or indirectly attaching cytoplasmic mRNPs to the cytoskeleton. Alternatively spliced transcript variants encoding the same protein have been found for this gene.

Sequence

Synthetic peptide located within the following region: EQLDQPISACCFNHNGNIFAYASSYDWSKGHEFYNPQKKNYIFLRNAAEE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service