Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AV39137

Sigma-Aldrich

Anti-TSC22D2 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-TSC22 domain family, member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

79 kDa

species reactivity

guinea pig, human, mouse, dog, rat, rabbit, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TSC22D2(9819)

Immunogen

Synthetic peptide directed towards the N terminal region of human TSC22D2

Biochem/physiol Actions

Transforming growth factor (TGF)-β-stimulated clone 22 domain family of leucine zippers is involved in cellular osmotic stress response. TSC22D isoforms are regulated by glucocorticoids and TGF-β and aid the renal cells to adapt to hypertonicity.

Sequence

Synthetic peptide located within the following region: CFQITSVTTAQVATSITEDTESLDDPDESRTEDVSSEIFDVSRATDYGPE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Liang et al.
International journal of oncology, 49(3), 1046-1056 (2016-08-31)
We previously identified TSC22D2 (transforming growth factor β-stimulated clone 22 domain family, member 2) as a novel cancer-associated gene in a rare multi-cancer family. However, its role in tumor development remains completely unknown. In this study, we found that TSC22D2 was
Diego F Fiol et al.
The FEBS journal, 274(1), 109-124 (2006-12-07)
We recently cloned a novel osmotic stress transcription factor 1 (OSTF1) from gills of euryhaline tilapia (Oreochromis mossambicus) and demonstrated that acute hyperosmotic stress transiently increases OSTF1 mRNA and protein abundance [Fiol DF, Kültz D (2005) Proc Natl Acad Sci

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service