Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV38768

Sigma-Aldrich

Anti-DPF2 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-D4, Zinc and double PHD fingers family 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

44 kDa

species reactivity

bovine, rat, guinea pig, horse, rabbit, goat, dog, human, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... DPF2(5977)

Immunogen

Synthetic peptide directed towards the N terminal region of human DPF2

Biochem/physiol Actions

DPF2 is a d4 domain-containing transcription factor that is essential for apoptotic response in the absence of survival factors. It associates with histone H3 and HDAC1 and acts as nuclear-selective co-repressor of estrogen-related receptor alpha.

Sequence

Synthetic peptide located within the following region: MAAVVENVVKLLGEQYYKDAMEQCHNYNARLCAERSVRLPFLDSQTGVAQ

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T G Gabig et al.
The Journal of biological chemistry, 269(47), 29515-29519 (1994-11-25)
To identify genes mediating programmed cell death triggered by interleukin 3 (IL-3)-deprivation of myeloid cells, the IL-3-dependent murine myeloid cell line FDCP-1 was used to screen a mammalian cell expression library for cDNAs that would promote survival following withdrawal of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service