Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV35579

Sigma-Aldrich

Anti-CACNB1 (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CAB1, Anti-CACNLB1, Anti-CCHLB1, Anti-Calcium channel, voltage-dependent, β 1 subunit, Anti-MGC41896

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

58 kDa

species reactivity

guinea pig, rabbit, bovine, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CACNB1(782)

Immunogen

Synthetic peptide directed towards the middle region of human CACNB1

Biochem/physiol Actions

CACNB1 belongs to the calcium channel b subunit family. It has important roles in the modulation of calcium channels and calcium current, as well as in voltage-dependent activation and inactivation of calcium channels.

Sequence

Synthetic peptide located within the following region: TRRPTPPASGNEMTNLAFELDPLELEEEEAELGEQSGSAKTSVSSVTTPP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Hogan et al.
Neuroscience letters, 277(2), 111-114 (2000-01-07)
The structure of the gene encoding the human brain beta1 subunit of voltage dependent calcium channels (CACNB1) was determined by comparison of its genomic sequence with beta1 cDNA sequence. CACNB1 is distributed over 25 kb and contains 13 exons. Alternative
Wanchana Jangsangthong et al.
Pflugers Archiv : European journal of physiology, 459(3), 399-411 (2009-10-13)
Voltage-dependent calcium channel (Ca(v)) pores are modulated by cytosolic beta subunits. Four beta-subunit genes and their splice variants offer a wide structural array for tissue- or disease-specific biophysical gating phenotypes. For instance, the length of the N terminus of beta(2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service