Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

AV35097

Sigma-Aldrich

Anti-KCNN3 (AB1) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

81 kDa

species reactivity

mouse, dog, rabbit, human, guinea pig, rat, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNN3(3782)

General description

KCNN3 (SK3) codes for a voltage-independent, calcium-activated channel integral membrane protein. It may be involved in the modulation of neuronal excitability. KCNN3 has been linked to anorexia nervosa risk and atrial fibrillation.
Rabbit Anti-KCNN3 antibody recognizes canine, human, mouse, rat, pig, and bovine KCNN3.

Immunogen

Synthetic peptide directed towards the C terminal region of human KCNN3

Application

Rabbit Anti-KCNN3 antibody is suitable for western blot assays at 2 μg/ml.

Biochem/physiol Actions

Action potentials in vertebrate neurons are followed by an afterhyperpolarization (AHP) that may persist for several seconds and may have profound consequences for the firing pattern of the neuron. Each component of the AHP is kinetically distinct and is mediated by different calcium-activated potassium channels. KCNN3, a member of the KCNN family of potassium channel genes, encodes a protein that is activated before membrane hyperpolarization and is thought to regulate neuronal excitability by contributing to the slow component of synaptic AHP. The encoded protein is an integral membrane protein that forms a voltage-independent calcium-activated channel with three other calmodulin-binding subunits.

Sequence

Synthetic peptide located within the following region: ITELNDRSEDLEKQIGSLESKLEHLTASFNSLPLLIADTLRQQQQQLLSA

Physical form

 Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maya Koronyo-Hamaoui et al.
American journal of medical genetics. Part B, Neuropsychiatric genetics : the official publication of the International Society of Psychiatric Genetics, 131B(1), 76-80 (2004-09-25)
The human small-conductance Ca(2+)-activated potassium channel gene KCNN3 has been involved in mechanisms underlying neuronal function and plasticity. A multiallelic CAG repeat polymorphism within the KCNN3 has been associated with schizophrenia and bipolar disorder. We have previously reported in a
Patrick T Ellinor et al.
Nature genetics, 42(3), 240-244 (2010-02-23)
Atrial fibrillation (AF) is the most common sustained arrhythmia. Previous studies have identified several genetic loci associated with typical AF. We sought to identify common genetic variants underlying lone AF. This condition affects a subset of individuals without overt heart

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service