Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV34677

Sigma-Aldrich

Anti-KLHL14 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Kelch-like 14 (Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

71 kDa

species reactivity

guinea pig, bovine, human, dog, rat, rabbit, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KLHL14(57565)

General description

KLHL14 is a protein member of the kelch-like family.
Rabbit anti-KLHL14 antibody recognizes chicken, canine, bovine, human, mouse, and rat KLHL14.

The previously assigned protein identifier Q8WU41 has been merged into Q9P2G3. Full details can be found on the UniProt database.

Immunogen

Synthetic peptide directed towards the N terminal region of human KLHL14

Application

Rabbit anti-KLHL14 antibody is suitable for western blot assays and for IHC assays at 4-8 μg/ml.

Biochem/physiol Actions

KLHL14 is a member of the KLHL family.

Sequence

Synthetic peptide located within the following region: MSRSGDRTSTFDPSHSDNLLHGLNLLWRKQLFCDVTLTAQGQQFHCHKAV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peng Hou et al.
Endocrine-related cancer, 18(6), 687-697 (2011-09-23)
The BRAF V600E mutation plays an important role in the tumorigenesis of papillary thyroid cancer (PTC). To explore an epigenetic mechanism involved in this process, we performed a genome-wide DNA methylation analysis using a methylated CpG island amplification (MCA)/CpG island
Peng Hou et al.
Cell cycle (Georgetown, Tex.), 11(2), 286-295 (2011-12-23)
Although BRAF(V600E) is well known to play an important role in the tumorigenesis of melanoma, its molecular mechanism, particularly the epigenetic aspect, has been incompletely understood. Here, we investigated the role of BRAF(V600E) signaling in altering gene methylation in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service