Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV34622

Sigma-Aldrich

Anti-KCNIP4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Kv channel interacting protein 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

29 kDa

species reactivity

mouse, rat, rabbit, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... KCNIP4(80333)

General description

KCNIP4 is a protein member of the voltage-gated potassium (Kv) channel-interacting family. This protein is known to associate with calcium and presenilin. KCNIP4 has been implicated as a candidate gene for renal cancer, personality and attention-deficit/hyperactivity disorders (PD and ADHD), and asthma.
Rabbit Anti-KCNIP4 antibody recognizes human, mouse, rat, bovine, and canine KCNIP4.

Immunogen

Synthetic peptide directed towards the N terminal region of human KCNIP4

Application

Rabbit Anti-KCNIP4 antibody is suitable for western blot (1.0 μg/ml) and IHC (4-8 μg/ml) applications.

Biochem/physiol Actions

KCNIP4 encodes a member of the family of voltage-gated potassium (Kv) channel-interacting proteins (KCNIPs), which belong to the recoverin branch of the EF-hand superfamily. Members of the KCNIP family are small calcium binding proteins. They all have EF-hand-like domains, and differ from each other in the N-terminus. They are integral subunit components of native Kv4 channel complexes. They may regulate A-type currents, and hence neuronal excitability, in response to changes in intracellular calcium. This protein member also interacts with presenilin.

Sequence

Synthetic peptide located within the following region: NSTKRSIKERLMKLLPCSAAKTSSPAIQNSVEDELEMATVRHRPEALELL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lena Weißflog et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 23(6), 436-447 (2012-09-18)
Attention-deficit/hyperactivity disorder (ADHD) is a neurodevelopmental disorder in children with striking persistence into adulthood and a high co-morbidity with other psychiatric disorders, including personality disorders (PD). The 4p15.31 region was shown to be associated with ADHD in several genome wide
Anita Bonne et al.
Cancer genetics and cytogenetics, 179(1), 11-18 (2007-11-06)
Our group and others had previously developed a high throughput procedure to map translocation breakpoints using chromosome flow sorting in conjunction with microarray-based comparative genomic hybridization (arrayCGH). Here we applied both conventional positional cloning and integrated arrayCGH procedures to the
Blanca E Himes et al.
PloS one, 8(2), e56179-e56179 (2013-03-05)
Asthma is a common chronic respiratory disease characterized by airway hyperresponsiveness (AHR). The genetics of asthma have been widely studied in mouse and human, and homologous genomic regions have been associated with mouse AHR and human asthma-related phenotypes. Our goal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service