Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV34548

Sigma-Aldrich

Anti-TRIM17 (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Tripartite motif-containing 17

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

54 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRIM17(51127)

General description

TRIM17 (Terf) is an E3 ubiquitin ligase that mediates Mcl-1 degradation and initiates apoptosis in neurons. Terf can also facilitate the degradation of ZWINT (a kinetochore protein) and decrease the proliferation of MCF7 breast cancer cells.
Rabbit Anti-TRIM17 antibody recognizes human TRIM17.

Immunogen

Synthetic peptide directed towards the C terminal region of human TRIM17

Application

Rabbit Anti-TRIM17 (AB2) antibody is suitable for western blot applications at a concentration of 1.25 μg/ml.

Biochem/physiol Actions

TRIM17 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. The protein is expressed almost exclusively in the testis, but its function is unknown. Multiple alternatively spliced transcript variants have been found for this gene.

Sequence

Synthetic peptide located within the following region: PKCPENGFWVVQLSKGTKYLSTFSALTPVMLMEPPSHMGIFLDFEAGEVS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroshi Endo et al.
Journal of biochemistry, 151(2), 139-144 (2011-10-26)
Terf/TRIM17 is a tripartite motif protein that has been originally isolated from testis. Terf has been characterized to exhibit an E3 ubiquitin ligase activity and to undergo self-ubiquitination. The cellular function of terf and its substrates, however, remain elusive. In
I Lassot et al.
Cell death and differentiation, 17(12), 1928-1941 (2010-06-19)
Accumulating data indicate that the ubiquitin-proteasome system controls apoptosis by regulating the level and the function of key regulatory proteins. In this study, we identified Trim17, a member of the TRIM/RBCC protein family, as one of the critical E3 ubiquitin
M M Magiera et al.
Cell death and differentiation, 20(2), 281-292 (2012-09-15)
Short-term proteasome inhibition has been shown to prevent neuronal apoptosis. However, the key pro-survival proteins that must be degraded for triggering neuronal death are mostly unknown. Here, we show that Mcl-1, an anti-apoptotic Bcl-2 family member, is degraded by the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service