Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV32707

Sigma-Aldrich

Anti-MyBL1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-A-MYB, Anti-AMYB, Anti-MGC120059, Anti-MGC120061, Anti-v-Myb myeloblastosis viral oncogene homolog (avian)-like 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

49 kDa

species reactivity

rabbit, mouse, horse, dog, rat, human, guinea pig, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYBL1(4603)

General description

Rabbit polyclonal anti-MyBL1 antibody reacts with chicken, canine, human, mouse, and rat v-Myb myeloblastosis viral oncogene homolog (avian)-like 1 transcription factors.
v-Myb myeloblastosis viral oncogene homolog (avian)-like 1 (MyBL1) which is expressed predominantly as a tissue-specific transcription factor in spermatocytes and breast epithelial cells is a male-specific regulator of several crucial meiotic processes. MYBL1 is a master regulator of meiotic genes that are involved in multiple processes in spermatocytes, particularly those required for cell cycle progression through pachynema. MYBL1 directs germ cell-specific activation via the CRE site of certain genes.

Immunogen

Synthetic peptide directed towards the middle region of human MYBL1

Application

Rabbit Anti-MyBL1 antibody can be used for IHC (4-8μg/ml) and western blot (2.5μg/ml) applications.
Rabbit polyclonal anti-MyBL1 antibody is used to tag v-Myb myeloblastosis viral oncogene homolog (avian)-like 1 for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of v-Myb myeloblastosis viral oncogene homolog (avian)-like 1 in spermatocyte development.

Biochem/physiol Actions

MYBL1 is strong transcriptional activator; DNA-binding protein that specifically recognize the sequence 5′-YAAC[GT]G-3′. It could have a role in the proliferation and/or differentiation of neurogenic, spermatogenic and B-lymphoid cells.

Sequence

Synthetic peptide located within the following region: PRTPTPFKNALAAQEKKYGPLKIVSQPLAFLEEDIREVLKEETGTDLFLK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service