Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

AV31874

Sigma-Aldrich

Anti-ZBTB32 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Zinc finger and BTB domain containing 32

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

33 kDa

species reactivity

rat, human, bovine, guinea pig, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZBTB32(27033)

General description

ZBTB32 is a zinc-finger protein that can repress the expression of CIITA and MHC II genes during B cell differentiation.
Rabbit Anti-ZBTB32 recognizes bovine, mouse, canine, and human ZBTB32.

Immunogen

Synthetic peptide directed towards the N terminal region of human ZBTB32

Application

Rabbit Anti-ZBTB32 antibody can be used for western blot applications at a concentration of 0.5μg/ml.

Biochem/physiol Actions

ZBTB32 may play an essential role during the proliferative stages of primitive hematopoietic progenitors, possibly acting in concert with (a subset of) the Fanconi anemia proteins. This gene can also interact with GATA-2 and can modify GATA-2 transactivation capacity.

Sequence

Synthetic peptide located within the following region: PIRLPSPYGSDRLVQLAARLRPALCDTLITVGSQEFPAHSLVLAGVSQQL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye Suk Yoon et al.
Journal of immunology (Baltimore, Md. : 1950), 189(5), 2393-2403 (2012-08-02)
CIITA and MHC class II expression is silenced during the differentiation of B cells to plasma cells. When B cell differentiation is carried out ex vivo, CIITA silencing occurs rapidly, but the factors contributing to this event are not known.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service