Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV31652

Sigma-Aldrich

Anti-ESR2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Estrogen receptor 2 (ER β)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

59 kDa

species reactivity

mouse, human, pig, bovine, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ESR2(2100)

General description

ESR2 is a nuclear transcription factor that belongs to the estrogen receptor family. ESR2 polymorphisms have been linked to anorexia nervosa, blood pressure, breast cancer and endometrial tumors.
Rabbit Anti-ESR2 antibody recognizes canine, human, mouse, rat, pig, and bovine ESR2.

Immunogen

Synthetic peptide directed towards the N terminal region of human ESR2

Application

Rabbit Anti-ESR2 antibody can be used for western blot applications at a dilution of 1μg/ml.

Biochem/physiol Actions

ESR2 is a member of the family of estrogen receptors and superfamily of nuclear receptor transcription factors. The gene product contains an N-terminal DNA binding domain and C-terminal ligand binding domain and is localized to the nucleus, cytoplasm, and mitochondria. Upon binding to 17beta-estradiol or related ligands, the encoded protein forms homo- or hetero-dimers that interact with specific DNA sequences to activate transcription. Some isoforms dominantly inhibit the activity of other estrogen receptor family members. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been fully characterized.

Sequence

Synthetic peptide located within the following region: TPGHLSPLVVHRQLSHLYAEPQKSPWCEARSLEHTLPVNRETLKRKVSGN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paula Maguire et al.
Breast cancer research and treatment, 94(2), 145-152 (2005-11-02)
Estrogen is involved in both normal mammary development and in breast carcinogenesis. A family history of disease and exposure to estrogen are major risk factors for developing breast cancer. Estrogen exerts its biological effects through binding to the estrogen receptors
Veronica Wendy Setiawan et al.
Cancer causes & control : CCC, 15(6), 627-633 (2004-07-29)
We hypothesized that variations in the ESR2 gene may influence estrogen exposure in the uterus and thus influence endometrial cancer risk. We validated and screened for variants in the ESR2 gene and examined whether they are associated with endometrial cancer
H Eastwood et al.
Molecular psychiatry, 7(1), 86-89 (2002-01-23)
There is significant evidence for genetic factors in the susceptibility to anorexia nervosa (AN). Previously genetic variation in the estrogen receptor 2 gene (ESR2) has been studied, however no strong evidence of association with AN has been found. In the
S Ogawa et al.
Journal of human genetics, 45(6), 327-330 (2001-02-24)
We investigated the association between a dinucleotide (cytosine-adenine; CA) repeat polymorphism located in the flanking region of the human estrogen receptor beta (ESR2) gene and systemic blood pressure in 187 healthy postmenopausal Japanese women. The genotype was classified as "A"

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service