Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AV31369

Sigma-Aldrich

Anti-KLF9 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Kruppel-like factor 9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

27 kDa

species reactivity

rat, rabbit, horse, mouse, human, bovine, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KLF9(687)

Related Categories

General description

Kruppel-like factor 9 (KLF9) is a transcription factor that regulates several functions such as central nervous systems (CNS) development, villus cell movement, intestinal cell proliferation, and PPARγ-mediated adipocyte differentiation. Furthermore, KLF9 also regulates the differentiation, adhesion and growth of endometrial cells and has been implicated in endometrial carcinoma.
Rabbit Anti-KLF9 antibody recognizes pig, bovine, human, mouse, and rat KLF9.

Immunogen

Synthetic peptide directed towards the N-terminal region of Human KLF9

Application

Rabbit Anti-KLF9 antibody is suitable for use in western blot (1.0μg/ml) and IHC (4-8μg/ml) applications.

Biochem/physiol Actions

KLF9 is a transcription factor that binds to GC box elements located in the promoter. Binding of the encoded protein to a single GC box inhibits mRNA expression while binding to tandemly repeated GC box elements activates transcription.

Sequence

Synthetic peptide located within the following region: LPEREVTKEHGDPGDTWKDYCTLVTIAKSLLDLNKYRPIQTPSVCSDSLE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Pei et al.
Cell death and differentiation, 18(2), 315-327 (2010-08-21)
Krüppel-like factors (KLFs) as a family of zinc-finger transcription factors involve in the regulation of many physiological processes. In these studies, KLF9 was characterized for its role in adipogenesis. The expression of KLF9 was markedly upregulated during the middle stage
Frank A Simmen et al.
Reproductive biology and endocrinology : RB&E, 6, 41-41 (2008-09-12)
Krüppel-like factor 9 (KLF9) is a transcriptional regulator of uterine endometrial cell proliferation, adhesion and differentiation; processes essential for pregnancy success and which are subverted during tumorigenesis. The network of endometrial genes controlled by KLF9 is largely unknown. Over-expression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service