Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV30074

Sigma-Aldrich

Anti-EEA1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Early endosome antigen 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

162 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EEA1(8411)

General description

Early Endosome Antigen1 (EEA1) is a conserved protein that regulates vesicular transport of proteins in early endosomes. Rabbit Anti-EEA1 antibody recognizes mouse and human EEA1.

Immunogen

Synthetic peptide directed towards the N terminal region of human EEA1

Application

Rabbit Anti-EEA1 antibody can be used for western blot (0.6μg/ml) applications.

Biochem/physiol Actions

EEA1 is involved in neuronal synaptic vesicle function and axonal transport and growth. EEA1 may undergo calcium-dependent conformational changes that are required for binding to SNAP-25.

Sequence

Synthetic peptide located within the following region: LTENLLKKEQDYTKLEEKHNEESVSKKNIQATLHQKDLDCQQLQSRLSAS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F T Mu et al.
The Journal of biological chemistry, 270(22), 13503-13511 (1995-06-02)
Early endosomes are cellular compartments receiving endocytosed material and sorting them for vesicular transport to late endosomes and lysosomes or for recycling to the plasma membrane. We have cloned a human cDNA encoding an evolutionarily conserved 180-kDa protein on early
Sandra Kissing et al.
The Journal of biological chemistry, 290(22), 14166-14180 (2015-04-24)
The vacuolar H(+)-ATPase (v-ATPase) complex is instrumental in establishing and maintaining acidification of some cellular compartments, thereby ensuring their functionality. Recently it has been proposed that the transmembrane V0 sector of v-ATPase and its a-subunits promote membrane fusion in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service