Skip to Content
MilliporeSigma

ADAP05-EP

Adapter for all Eppendorf® 1.5/2.0 mL Rotors

holds 1 x 0.5 mL tubes, pack of 6 ea

Synonym(s):

bucket adapter, rotor adapter, tube adapter

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41103911
Compatibility:
for use with Rotors 1.5/2.0 mL
Manufacturer/tradename:
Eppendorf® 022636227 (NA)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


feature

holds 1 x 0.5 mL tubes

packaging

pack of 6 ea

manufacturer/tradename

Eppendorf® 022636227 (NA)

parameter

18,400 × g max. RCF

technique(s)

centrifugation: suitable

compatibility

for use with Rotors 1.5/2.0 mL

General description

Six adapters, each holding 1 PCR tube (0.2 mL, maximum diameter 6 mm). They can be used in all Eppendorf rotors holding 1.5/2.0 mL tubes.

Application

Adaption of various tube sizes to facilitate centrifugation

Features and Benefits

  • Adapts 0.2 mLPCR tubes to fit 1.5/2.0 mL tube rotors
  • Fits all Eppendorf rotos holding 1.5/2.0 mL tubes

Legal Information

Eppendorf is a registered trademark of Eppendorf SE


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library