Skip to Content
MilliporeSigma

A9376

Adenosine 2′:3′-cyclic monophosphate sodium salt

≥93%, synthetic (organic), powder

Synonym(s):

2′,3′-cAMP sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H11N5O6PNa
CAS Number:
Molecular Weight:
351.19
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.51
EC Number:
253-328-3
MDL number:
Assay:
≥93%
Biological source:
synthetic (organic)
Form:
powder
Solubility:
water: 50 mg/mL, clear, colorless
Storage temp.:
−20°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Adenosine 2′:3′-cyclic monophosphate sodium salt, ≥93%

biological source

synthetic (organic)

Quality Segment

assay

≥93%

form

powder

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Na+].Nc1ncnc2n(cnc12)[C@@H]3O[C@H](CO)[C@H]4OP([O-])(=O)O[C@@H]34

InChI

1S/C10H12N5O6P.Na/c11-8-5-9(13-2-12-8)15(3-14-5)10-7-6(4(1-16)19-10)20-22(17,18)21-7;/h2-4,6-7,10,16H,1H2,(H,17,18)(H2,11,12,13);/q;+1/p-1/t4-,6-,7-,10-;/m1./s1

InChI key

VSDSIACSNXHGOV-MCDZGGTQSA-M

Application

Adenosine 2′:3′-cyclic monophosphate sodium salt has been used in lysis buffer for affinity purification and also in microscale thermophoresis measurements. It has also been used in electrophysiological studies of Tenebrio tubules.

Biochem/physiol Actions

Adenosine 2′,3′-cyclic monophosphate (2′,3′-cAMP) is believed to serve as an extracellular source of adenosine. The release of extracellular 2′,3′-cAMP occurs in response to injury. 2′,3′-cAMP may be used to study the distribution and specificity of its degrading enzymes in the context of unique biological activities. 2′,3′-cAMP may also be used to study apoptosis induced at the level of mitochondrial permeability transition pores. 2′,3′-cAMP is converted into 2′-AMP and 3′-AMP which inhibit proliferation of preglomerular vascular smooth muscle cells and glomerular mesangial cells via A2B receptors.

Disclaimer

Caution: Do not confuse with the common adenosine 2′(3′)-monophosphate (mixed isomers).


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library