Skip to Content
MilliporeSigma

A9205

Bovine Serum Albumin solution

30% in saline, fatty acid free, aseptically filled

Synonym(s):

Albumin solution from bovine serum, BSA solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.27
MDL number:
eCl@ss:
32160409
Biological source:
bovine
Assay:
28-32% protein basis (biuret)
Form:
solution
Technique(s):
ELISA: suitable, electrophoresis: suitable, western blot: suitable
Impurities:
≤0.01% free fatty acid
Concentration:
28-32% protein (biuret), 30% in saline
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

Quality Segment

sterility

aseptically filled

assay

28-32% protein basis (biuret)

form

solution

purified by

cold ethanol fractionation

does not contain

stabilizer or preservative

origin

USA origin

concentration

28-32% protein (biuret), 30% in saline

free fatty acid content

≤0.01%

technique(s)

ELISA: suitable, electrophoresis: suitable, western blot: suitable

impurities

≤0.01% free fatty acid

UniProt accession no.

foreign activity

BT virus, none detected, VSV virus, none detected

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

General description

Bovine Serum Albumin (BSA) has three homologous domains, each subdivided it two domain (A and B). It is a α-helical globular protein and shares structural features with human serum albumin.

Application

Bovine Serum Albumin solution has been used:
  • as Dulbecco′s modified Eagle′s medium supplement for culturing HepG2 human liver cancer cells
  • as a component of hepatocyte recovery medium
  • as a component of fusion and activation buffers for in vitro reconstructed sheep embryos

Biochem/physiol Actions

Bovine Serum Albumin (BSA) is the major protein present in blood and has calcium binding sites and regulates metal homeostasis. It is a crucial component of the cell culture medium and promotes embryonic stem cells (hESC) differentiation. Due to its high solubility, low cost and purity it is widely used in cosmetic and pharmaceutical industry. BSA is used as a blocking agent used in enzyme linked immunosorbent assay (ELISA) for preventing non-specific binding of antigens and antibodies to the microtiter plates.

Preparation Note

Initial fractionation by cold alcohol precipitation


Still not finding the right product?

Explore all of our products under Bovine Serum Albumin solution


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library