Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A9111

Sigma-Aldrich

(±)-AMPA hydrobromide

≥98% (HPLC)

Synonym(s):

(±)-α-Amino-3-hydroxy-5-methylisoxazole-4-propionic acid hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2O4 · HBr
CAS Number:
Molecular Weight:
267.08
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

off-white to light tan

solubility

H2O: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

Br.Cc1onc(O)c1CC(N)C(O)=O

InChI

1S/C7H10N2O4.BrH/c1-3-4(6(10)9-13-3)2-5(8)7(11)12;/h5H,2,8H2,1H3,(H,9,10)(H,11,12);1H

InChI key

KUAHVIUZGLGASU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

(±)-AMPA is a potent excitatory amino acid that interacts selectively with central AMPA/kainate receptors.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to ion channel family glutamate receptors for your research needs.

We offers many products related to ion channel family glutamate receptors for your research needs.

We offers many products related to ion channel family glutamate receptors for your research needs.

We offers many products related to ion channel family glutamate receptors for your research needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service